Assay Details
Gene Symbol: PLPPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970142.1 | XP_027825943.1 | 7 | 1130 | 113 | |
XM_027970143.1 | XP_027825944.1 | 7 | 1149 | 113 | ||
XM_027970144.1 | XP_027825945.1 | 8 | 1058 | 113 | ||
XM_027970145.1 | XP_027825946.1 | 7 | 1898 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
