Assay Details
Gene Symbol: ZUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011177.4 | XP_004011226.2 | 8 - 9 | 1782 | 112 | |
XM_012182530.3 | XP_012037920.2 | 8 - 9 | 1730 | 112 | ||
XM_027972349.1 | XP_027828150.1 | 8 - 9 | 1817 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
