Assay Details
Gene Symbol: LOC114115942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972279.1 | XP_027828080.1 | 2 | 878 | 53 |
Gene Symbol: GAS2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956711.1 | XP_027812512.1 | 1 - 2 | 1501 | 53 | |
XM_027956712.1 | XP_027812513.1 | 2 - 3 | 826 | 53 | ||
XM_027956713.1 | XP_027812514.1 | 2 - 3 | 881 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
