Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092979.2 | XP_014948465.2 | 19 - 20 | 2112 | 88 | |
XM_015092980.2 | XP_014948466.2 | 18 - 19 | 2035 | 88 | ||
XM_027964118.1 | XP_027819919.1 | 18 - 19 | 2295 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
