Assay Details
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103531.3 | XP_011958921.1 | 5 - 6 | 2287 | 74 | |
XM_027960594.1 | XP_027816395.1 | 5 - 6 | 2048 | 74 | ||
XM_027960603.1 | XP_027816404.1 | 4 - 5 | 2115 | 74 | ||
XM_027960605.1 | XP_027816406.1 | 5 - 6 | 2287 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
