Assay Details
Gene Symbol: EXOC3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190547.3 | XP_012045937.2 | 10 - 11 | 2155 | 94 | |
XM_027978471.1 | XP_027834272.1 | 10 - 11 | 2155 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
