Assay Details
Gene Symbol: PLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016984.4 | XP_004017033.1 | 3 - 4 | 849 | 94 | |
XM_012096933.3 | XP_011952323.2 | 3 - 4 | 838 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
