Assay Details
Gene Symbol: RPL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103959.3 | XP_011959349.2 | 8 - 9 | 1215 | 55 | |
XM_027961461.1 | XP_027817262.1 | 8 - 9 | 1201 | 55 | ||
XM_027961462.1 | XP_027817263.1 | 8 - 9 | 1254 | 55 | ||
XM_027961463.1 | XP_027817264.1 | 8 - 9 | 1240 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
