Assay Details
Gene Symbol: E2F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973255.1 | XP_027829056.1 | 5 - 6 | 574 | 103 | |
XM_027973256.1 | XP_027829057.1 | 5 - 6 | 574 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
