Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968079.1 | XP_027823880.1 | 2 - 3 | 873 | 66 | |
XM_027968080.1 | XP_027823881.1 | 2 - 3 | 954 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
