Assay Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011320.4 | XP_004011369.1 | 1 - 2 | 170 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
