Assay Details
Gene Symbol: KIF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018499.4 | XP_004018548.2 | 20 - 21 | 2793 | 86 | |
XM_012100140.3 | XP_011955530.2 | 20 - 21 | 2790 | 86 | ||
XM_027957719.1 | XP_027813520.1 | 19 - 20 | 2547 | 86 |
Gene Symbol: LOC114108744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963798.1 | XP_027819599.1 | 3 - 4 | 340 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
