Assay Details
Gene Symbol: KIF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018499.4 | XP_004018548.2 | 21 - 22 | 2898 | 69 | |
XM_012100140.3 | XP_011955530.2 | 21 - 22 | 2895 | 69 | ||
XM_027957719.1 | XP_027813520.1 | 20 - 21 | 2652 | 69 |
Gene Symbol: LOC114108744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963798.1 | XP_027819599.1 | 4 - 5 | 445 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
