Assay Details
Gene Symbol: ADPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015091860.2 | XP_014947346.2 | 5 - 6 | 816 | 71 | |
XM_027967823.1 | XP_027823624.1 | 5 - 6 | 2255 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
