Assay Details
Gene Symbol: SCIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185560.3 | XP_012040950.1 | 2 - 3 | 540 | 92 | |
XM_012185561.3 | XP_012040951.1 | 2 - 3 | 408 | 92 | ||
XM_027975082.1 | XP_027830883.1 | 2 - 3 | 542 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
