Assay Details
Gene Symbol: ZNF446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978941.1 | XP_027834742.1 | 2 - 3 | 406 | 87 | |
XM_027978942.1 | XP_027834743.1 | 2 - 3 | 564 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
