Assay Details
Gene Symbol: LOC101122940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175443.3 | XP_012030833.2 | 18 - 19 | 2324 | 95 | |
XM_012175444.3 | XP_012030834.2 | 18 - 19 | 2544 | 95 | ||
XM_012175446.3 | XP_012030836.2 | 18 - 19 | 2531 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
