Assay Details
Gene Symbol: LOC101122940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175443.3 | XP_012030833.2 | 29 - 30 | 3835 | 76 | |
XM_012175444.3 | XP_012030834.2 | 29 - 30 | 4055 | 76 | ||
XM_012175446.3 | XP_012030836.2 | 29 - 30 | 4042 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
