Assay Details
Gene Symbol: MMUT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018874.4 | XP_004018923.1 | 10 - 11 | 2000 | 78 | |
XM_004018875.4 | XP_004018924.1 | 10 - 11 | 1971 | 78 | ||
XM_012100805.3 | XP_011956195.1 | 10 - 11 | 1994 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
