Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972144.1 | XP_027827945.1 | 39 - 40 | 7119 | 127 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972144.1 | XP_027827945.1 | 39 - 40 | 7119 | 127 |
Set Membership: |
![]() ![]() |