Assay Details
Gene Symbol: PROCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012504.4 | XP_004012553.1 | 3 - 4 | 1396 | 82 | |
XM_012185400.3 | XP_012040790.1 | 4 - 5 | 1499 | 82 | ||
XM_012185401.3 | XP_012040791.1 | 2 - 3 | 1310 | 82 |
