Assay Details
Gene Symbol: HEXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013048.4 | XP_004013097.3 | 12 - 13 | 1364 | 109 | |
XM_027974313.1 | XP_027830114.1 | 12 - 13 | 1364 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
