Assay Details
Gene Symbol: HEXD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013048.4 | XP_004013097.3 | 13 - 14 | 1448 | 88 | |
XM_027974312.1 | XP_027830113.1 | 12 - 13 | 1346 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
