Assay Details
Gene Symbol: AOPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964236.1 | XP_027820037.1 | 9 - 10 | 2051 | 90 | |
XM_027964237.1 | XP_027820038.1 | 9 - 10 | 2051 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
