Assay Details
Gene Symbol: MFSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973415.1 | XP_027829216.1 | 1 - 2 | 1584 | 86 | |
XM_027973420.1 | XP_027829221.1 | 2 - 3 | 1306 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
