Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976600.1 | XP_027832401.1 | 16 - 17 | 2349 | 64 | |
XM_027976601.1 | XP_027832402.1 | 16 - 17 | 2347 | 64 | ||
XM_027976602.1 | XP_027832403.1 | 16 - 17 | 2348 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
