Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976600.1 | XP_027832401.1 | 2 - 3 | 442 | 110 | |
XM_027976601.1 | XP_027832402.1 | 2 - 3 | 440 | 110 | ||
XM_027976602.1 | XP_027832403.1 | 2 - 3 | 441 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
