Assay Details
Gene Symbol: LOC101115252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961640.1 | XP_027817441.1 | 11 - 12 | 1521 | 108 | |
XM_027961641.1 | XP_027817442.1 | 11 - 12 | 1332 | 108 | ||
XM_027961642.1 | XP_027817443.1 | 12 - 13 | 1384 | 108 | ||
XM_027961643.1 | XP_027817444.1 | 11 - 12 | 1274 | 108 | ||
XM_027961644.1 | XP_027817445.1 | 10 - 11 | 1313 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
