Assay Details
Gene Symbol: LOC101115252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961640.1 | XP_027817441.1 | 5 - 6 | 791 | 72 | |
XM_027961641.1 | XP_027817442.1 | 5 - 6 | 602 | 72 | ||
XM_027961642.1 | XP_027817443.1 | 6 - 7 | 654 | 72 | ||
XM_027961643.1 | XP_027817444.1 | 5 - 6 | 544 | 72 | ||
XM_027961644.1 | XP_027817445.1 | 4 - 5 | 583 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
