Assay Details
Gene Symbol: LOC101115252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961640.1 | XP_027817441.1 | 7 - 8 | 1008 | 74 | |
XM_027961641.1 | XP_027817442.1 | 7 - 8 | 819 | 74 | ||
XM_027961642.1 | XP_027817443.1 | 8 - 9 | 871 | 74 | ||
XM_027961643.1 | XP_027817444.1 | 7 - 8 | 761 | 74 | ||
XM_027961644.1 | XP_027817445.1 | 6 - 7 | 800 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
