Assay Details
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968173.1 | XP_027823974.1 | 4 - 5 | 411 | 104 | |
XM_027968174.1 | XP_027823975.1 | 4 - 5 | 411 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
