Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959398.1 | XP_027815199.1 | 11 - 12 | 4010 | 108 | |
XM_027959399.1 | XP_027815200.1 | 12 - 13 | 3926 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
