Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959398.1 | XP_027815199.1 | 5 - 6 | 3178 | 59 | |
XM_027959399.1 | XP_027815200.1 | 6 - 7 | 3094 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
