Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959398.1 | XP_027815199.1 | 7 - 8 | 3558 | 75 | |
XM_027959399.1 | XP_027815200.1 | 8 - 9 | 3474 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
