Assay Details
Gene Symbol: TMPRSS11F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970749.1 | XP_027826550.1 | 5 - 6 | 638 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
