Assay Details
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008002.4 | XP_004008051.1 | 2 - 3 | 391 | 108 | |
XM_027968754.1 | XP_027824555.1 | 2 - 3 | 285 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
