Assay Details
Gene Symbol: INTS6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022318.4 | XP_004022367.2 | 7 - 8 | 1303 | 72 | |
XM_004022319.4 | XP_004022368.2 | 7 - 8 | 1304 | 72 | ||
XM_015105013.2 | XP_014960499.2 | 7 - 8 | 1303 | 72 | ||
XM_027963641.1 | XP_027819442.1 | 7 - 8 | 1303 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
