Assay Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103831.3 | XP_011959221.3 | 1 - 2 | 1575 | 96 | |
XM_027960877.1 | XP_027816678.1 | 2 - 3 | 1361 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
