Assay Details
Gene Symbol: TCFL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188858.3 | XP_012044248.2 | 5 - 6 | 1388 | 96 | |
XM_027976968.1 | XP_027832769.1 | 5 - 6 | 1385 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
