Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962502.1 | XP_027818303.1 | 2 - 3 | 853 | 91 | |
XM_027962503.1 | XP_027818304.1 | 2 - 3 | 853 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
