Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104640.2 | XP_014960126.1 | 4 - 5 | 865 | 84 | |
XM_015104641.2 | XP_014960127.1 | 3 - 4 | 346 | 84 | ||
XM_027962675.1 | XP_027818476.1 | 3 - 4 | 760 | 84 | ||
XM_027962676.1 | XP_027818477.1 | 4 - 5 | 422 | 84 | ||
XM_027962677.1 | XP_027818478.1 | 3 - 4 | 511 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
