Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013093.4 | XP_004013142.1 | 3 - 4 | 551 | 83 | |
XM_012186489.3 | XP_012041879.1 | 3 - 4 | 554 | 83 | ||
XM_027974216.1 | XP_027830017.1 | 3 - 4 | 716 | 83 | ||
XM_027974217.1 | XP_027830018.1 | 3 - 4 | 719 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
