Assay Details
Gene Symbol: SPINDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959293.1 | XP_027815094.1 | 1 - 2 | 2486 | 104 | |
XM_027959294.1 | XP_027815095.1 | 2 - 3 | 623 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
