Assay Details
Gene Symbol: RGS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976265.1 | XP_027832066.1 | 2 - 3 | 3249 | 76 | |
XM_027976266.1 | XP_027832067.1 | 3 - 4 | 2762 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
