Assay Details
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018540.3 | XP_004018589.1 | 3 - 4 | 378 | 95 | |
GenBank mRNA | AF190579.1 | - | 2 - 3 | 261 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
