Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959995.1 | XP_027815796.1 | 2 - 3 | 205 | 80 | |
XM_027959996.1 | XP_027815797.1 | 2 - 3 | 205 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
