Assay Details
Gene Symbol: RELB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100238.2 | XP_014955724.2 | 4 - 5 | 599 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
