Assay Details
Gene Symbol: PBXIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975818.1 | XP_027831619.1 | 7 - 8 | 923 | 56 | |
XM_027975824.1 | XP_027831625.1 | 7 - 8 | 920 | 56 | ||
XM_027975833.1 | XP_027831634.1 | 7 - 8 | 735 | 56 | ||
XM_027975842.1 | XP_027831643.1 | 5 - 6 | 545 | 56 | ||
XM_027975852.1 | XP_027831653.1 | 5 - 6 | 555 | 56 | ||
XM_027975857.1 | XP_027831658.1 | 5 - 6 | 592 | 56 | ||
XM_027975863.1 | XP_027831664.1 | 5 - 6 | 542 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
