Assay Details
Gene Symbol: MRTFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962081.1 | XP_027817882.1 | 13 - 14 | 1626 | 66 | |
XM_027962082.1 | XP_027817883.1 | 11 - 12 | 1391 | 66 | ||
XM_027962083.1 | XP_027817884.1 | 11 - 12 | 1535 | 66 | ||
XM_027962084.1 | XP_027817885.1 | 11 - 12 | 1366 | 66 | ||
XM_027962085.1 | XP_027817886.1 | 9 - 10 | 2635 | 66 | ||
XM_027962086.1 | XP_027817887.1 | 13 - 14 | 1626 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
