Assay Details
Gene Symbol: MRTFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962081.1 | XP_027817882.1 | 18 - 19 | 3122 | 97 | |
XM_027962082.1 | XP_027817883.1 | 16 - 17 | 2887 | 97 | ||
XM_027962083.1 | XP_027817884.1 | 16 - 17 | 3031 | 97 | ||
XM_027962084.1 | XP_027817885.1 | 16 - 17 | 2862 | 97 | ||
XM_027962085.1 | XP_027817886.1 | 14 - 15 | 4131 | 97 | ||
XM_027962086.1 | XP_027817887.1 | 18 - 19 | 2975 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
